Learn More
Medchemexpress LLC Penicillin-Streptomycin-Neomycin (PSN) | 100 ML
SDP
Supplier:  Medchemexpress LLC HYK0014100ML
Encompass_Preferred
MCE Penicillin-Streptomycin-Neomycin (PSN) (100x), Sterile is a liquid formulation used to prevent bacterial contamination in cell cultures. It is effective against both gram-positive and gram-negative bacteria. The formulation contains 5,000 units/mL of penicillin, 5 mg/mL of streptomycin, and 10 mg/mL of Neomycin in a 0.85% NaCl solution.
  • Prevents bacterial contamination of cell culture
  • Effective combined action against gram-positive and gram-negative bacteria
  • Liquid formulation in 0.85% NaCl solution
Catalog No. 50-004-31440