Learn More
Alkali Scientific Penicillin/Streptomycin Solution, 50 Milliliters
SDP
Supplier:  Alkali Scientific NIC001CP
Encompass_Preferred
Penicillin/Streptomycin Solution, 50 mL, is a commonly used antimicrobial solution for cell culture applications. This solution contains Penicillin, a beta-lactam antibiotic effective against gram-positive bacteria, and Streptomycin, an aminoglycoside that targets gram-negative bacteria. The combination of both antibiotics makes this solution ideal for preventing bacterial contamination in cell cultures, especially for primary cultures or those involving sensitive mammalian cells. The 50 mL liquid format is convenient for preparing working solutions that are added to culture media. This solution is widely used in research and biopharmaceutical laboratories to maintain sterile culture conditions and prevent bacterial overgrowth during cell growth and experimentation.
Catalog No. 50-006-61421