Learn More
Medchemexpress LLC Penicillin V-d5 | 1356837-87-0 | 99.5% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0975AS5MG
Encompass_Preferred
Penicillin V-d5 (Phenoxymethylpenicillin-d5) is the deuterium-labeled Penicillin V. Penicillin V (Phenoxymethylpenicillin) is an orally active antibiotic that inhibits the growth of Streptococci, C. difficile, and S. aureus. It can be used for research on otitis, sinusitis, pharyngitis, and tonsillitis. The deuterium labeling allows it to be used as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Orally active antibiotic
  • Inhibits growth of Streptococci, C. difficile, and S. aureus
  • Suitable for research on otitis, sinusitis, pharyngitis, and tonsillitis
  • Deuterium-labeled compound
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08798