Learn More
Medchemexpress LLC Penicillin V-d5 potassium (phenoxymethylpenicillin-d5 potassium salt) | 2699607-22-0 | 98.10% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0975S11MG
Encompass_Preferred
Penicillin V-d5 potassium is the deuterium labeled Penicillin V Potassium. Penicillin V Potassium (Phenoxymethylpenicillin potassium salt) is an orally active antibiotic that inhibits the growth of Streptococci, C. difficile, and S. aureus. It can be used for research on otitis, sinusitis, pharyngitis, and tonsillitis.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.
Catalog No. 50-004-08048