Learn More
Apexbio Technology LLC Penicillin V potassium salt 132-98-9 200mg
SDP
Supplier:  Apexbio Technology LLC C7123200
Encompass_Preferred
Penicillin V potassium salt (132-98-9) is a small-molecule inhibitor targeting bacterial cell wall peptidoglycan synthesis It is designed to inhibit this biosynthetic pathway thereby suppressing the growth of Gram-positive bacteria Penicillin V potassium salt exerts its biological activity primarily through inhibition of bacterial cell wall synthesis In microbial susceptibility assays Penicillin V potassium salt demonstrates inhibitory effects with IC50 values ranging from approximately 0 01 to 0 5 g/mL for sensitive bacterial strains Based on these pharmacological properties Penicillin V potassium salt holds research potential in studies investigating Gram-positive bacterial resistance mechanisms cell-wall targeting antibiotic actions and experimental models of upper respiratory tract infections including otitis media sinusitis pharyngitis and tonsillitis
Catalog No. 50-000-94714