Learn More
Cayman Chemical PenIcolInate A 5mg
SDP
Supplier:  Cayman Chemical 250135 MG
Encompass_Preferred
A fungal metabolite with diverse biological activities; exhibits antimalarial and antituberculosis activity (MICs = 3.25 and 12.5 mg/ml for P. falciparum and M. tuberculosis H37Ra, respectively); cytotoxic to MCF-7, KB, NCI-H187, and Vero cells (IC50s = 7.49, 18.43, 4.4, and 14.08 μM, respectively)
Catalog No. 50-293-4414