Learn More
Medchemexpress LLC Pennogenin | 507-89-1 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN77981MG
Encompass_Preferred
Pennogenin is a bioactive component isolated from *T. govanianum* rhizomes. It demonstrates a significant *in vitro* inhibitory effect on the release of Reactive Oxygen Species (ROS).
  • Exhibits significant oxidative burst suppressive activity.
  • Shows a significant *in vitro* inhibitory effect on ROS release from whole blood, with an IC50 of 5.0 μg/mL.
  • For research use only.
Catalog No. 50-004-18376