Learn More
Cayman Chemical PentadecafluoroctnoIc AcId 25g
SDP
Supplier:  Cayman Chemical 3723225 G
Encompass_Preferred
A PFAS; chronic administration leads to accumulation in the kidney, liver, lung, heart, testes, and brain in rats at 5 and 20 mg/kg per day; is toxic to rats, inducing weight loss, sensitivity to external stimuli, lethargy, and hepatic inflammation at 20 mg/kg per day; found in marine life including lake trout, salmon, and whitefish, and as an environmental contaminant
Catalog No. 50-291-4781