Learn More
Medchemexpress LLC ω-Pentadecalactone | 106-02-5 | MFCD00039667 | 99.7% | 240.38 | C15H28O2 | 500 G

Supplier: Medchemexpress LLC HYW035362500G
ω-Pentadecalactone is a macrocyclic lactone used as a fragrance ingredient in fine fragrances, cosmetics, toiletries, and some household products. It is a colorless to light-yellow solid below 32°C (liquid above 32°C), with formula C15H28O2, molecular weight 240.38, and reported purity of 99.71%.
- Used in fragrances for cosmetics, shampoos, soaps, and household cleaners.
- Reported high purity suitable for formulation or research use (99.71%).
- Physical state changes at ~32°C - consider handling and storage conditions.
- Storage stability information available for pure material and in solution.
- Available in multiple pack sizes up to 500 g to support formulation and scale needs.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.