Learn More
Medchemexpress LLC Pentadecanedioic acid | 1460-18-0 | 98.0% | 272.38 | 100 G

Supplier: Medchemexpress LLC HYW011636100G
Pentadecanedioic acid (1,13-Tridecanedicarboxylic acid) is an ester product intended for research use only. It is not sold to patients. This compound is characterized by its solid, white to off-white appearance and high purity.
- Suitable for research applications
- Appears as a solid
- White to off-white in color
- Store at room temperature for 3 years, or in solvent at -80°C for 2 years or -20°C for 1 year
- Soluble in DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.