Learn More
Medchemexpress LLC Pentadecanedioic acid | 1460-18-0 | MFCD00039534 | 98.0% | 272.38 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01163650G
Encompass_Preferred
Pentadecanedioic acid is an ester product, appearing as a white to off-white solid. It is soluble in DMSO at 100 mg/mL and is for research use only, not validated for medical applications.
  • High purity of 98.0%
  • Molecular weight of 272.38
  • Store product at room temperature for 3 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
  • 1H NMR spectrum consistent with structure
Catalog No. 50-004-55151