Learn More
Medchemexpress LLC Pentadecanoic acid | 1002-84-2 | 242.40 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00428310G
Encompass_Preferred
Pentadecylic acid is a saturated fatty acid with a 15-carbon backbone. It induces apoptosis in MCF-7/SC breast cancer stem-like cells by inhibiting the JAK2/STAT3 signaling pathway and suppresses their proliferation and migration. Pentadecanoic acid also induces mild glucose intolerance during pregnancy in C57BL6/J mice, partly mediated through activation of PPARα and MAPK signaling pathways, and promotes offspring growth and enhanced antioxidant capacity during lactation.
  • Induces apoptosis in MCF-7/SC breast cancer stem-like cells.
  • Inhibits the JAK2/STAT3 signaling pathway.
  • Suppresses proliferation and migration of breast cancer cells.
  • Induces mild glucose intolerance during pregnancy in C57BL6/J mice.
  • Mediated through activation of PPARα and MAPK signaling pathways.
  • Promotes offspring growth and enhanced antioxidant capacity during lactation.
Catalog No. 50-004-47992