Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentadecanoic acid | 1002-84-2 | 242.40 | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHYW004283R

Encompass_Preferred

Pentadecanoic acid (Pentadecylic acid) is a saturated fatty acid with a 15-carbon backbone. It can induce apoptosis in breast cancer stem-like cells by inhibiting the JAK2/STAT3 signaling pathway and suppresses their proliferation and migration. It can also induce mild glucose intolerance during pregnancy, partly mediated through activation of PPARα and MAPK signaling pathways, and promoted offspring growth and enhanced antioxidant capacity during lactation.

  • Molecular weight: 242.40
  • Formula: C15H30O2
  • CAS number: 1002-84-2
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 99.92%
  • Structure classification: Ketones, aldehydes, acids
  • Initial source: Microorganisms
  • Endogenous metabolite
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-49966


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.