Learn More
Medchemexpress LLC Pentadecanoic acid | 1002-84-2 | 242.40 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00428350G
Encompass_Preferred
Pentadecylic acid is a saturated fatty acid with a 15-carbon backbone, utilized primarily for research purposes. It is not sold to patients.
  • Induces apoptosis and suppresses proliferation and migration in MCF-7/SC breast cancer stem-like cells.
  • Inhibits the JAK2/STAT3 signaling pathway.
  • Reduces MCF-7/SC cell viability and causes accumulation in Sub-G1 phase.
  • Induces mild glucose intolerance during pregnancy in C57BL6/J mouse models.
  • Promotes offspring growth and enhances antioxidant capacity during lactation.
Catalog No. 50-004-47151