Learn More
Medchemexpress LLC Pentadecanoic acid | 1002-84-2 | 242.40 | 500 MG

Supplier: Medchemexpress LLC HYW004283R500MG
Pentadecanoic acid is an analytical standard used in various research and analytical applications, including HPLC, GC, and MS. It is an endogenous metabolite and originates from microorganisms. This product is a white to off-white solid.
- Used as an analytical standard
- Suitable for HPLC, GC, and MS applications
- Endogenous metabolite
- Originates from microorganisms
- White to off-white solid
- High purity
- Molecular weight 242.40
- CAS number 1002-84-2
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.