Learn More
Medchemexpress LLC Pentadecanoic acid | 1002-84-2 | MFCD00002745 | 242.40 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00428310MM/1ML
Encompass_Preferred
Pentadecanoic acid is a saturated fatty acid with a 15-carbon backbone. It is a human endogenous metabolite.
  • Induces apoptosis in MCF-7/SC breast cancer stem-like cells by inhibiting the JAK2/STAT3 signaling pathway.
  • Suppresses proliferation and migration of MCF-7/SC cells.
  • Reduces cell viability in a dose- and time-dependent manner.
  • Reduces the ability of cells to penetrate Matrigel.
  • Causes accumulation of cells in the Sub-G1 phase, suggesting cell cycle arrest.
  • Induces mild glucose intolerance during pregnancy in C57BL6/J mice.
  • Promotes offspring growth and enhances antioxidant capacity during lactation.
Catalog No. 50-004-47150