Learn More
Medchemexpress LLC Pentadecanoic acid-d29 | 130348-95-7 | 271.58 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW004283S1MG
Encompass_Preferred
Pentadecanoic acid-d29 is the deuterium labeled pentadecanoic acid. Pentadecylic acid is a saturated fatty acid with a 15-carbon backbone. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled
  • Saturated fatty acid
  • Can be used as a tracer
  • Internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
Catalog No. 50-004-49134