Learn More
Cayman Chemical PentadecnoIc AcId-d29 10mg
SDP
Supplier:  Cayman Chemical 2808110 MG
Encompass_Preferred
An internal standard for the quantification of pentadecanoic acid by GC- or LC-MS
Catalog No. 50-293-8356