Learn More
Cayman Chemical PentadecnoIc AcId-d29 5mg
SDP
Supplier:  Cayman Chemical 280815 MG
Encompass_Preferred
An internal standard for the quantification of pentadecanoic acid by GC- or LC-MS
Catalog No. 50-294-0775