Learn More
Medchemexpress LLC Pentaerythritol tetra(3-mercaptopropionate) | 7575-23-7 | MFCD00022104 | 95.0% | 488.66 g/mol | C17H28O8S4 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW250309500G
Encompass_Preferred
Pentaerythritol tetra(3-mercaptopropionate) is a thiol-functional tetravalent ester used as a crosslinker and stabilizer in polymer, coating, adhesive, and specialty chemical formulations. It contains four mercaptopropionate groups on a pentaerythritol core, providing multiple reactive thiol sites for curing and formulation development.
  • Provides four reactive thiol sites for efficient crosslinking.
  • Enables thiol-ene and radical curing pathways.
  • Improves mechanical strength and thermal stability of cured materials.
  • Suitable for polymers, coatings, adhesives, and specialty formulations.
  • High purity for consistent performance in research and industrial use.
Catalog No. 50-002-81818