Learn More
Medchemexpress LLC Pentaerythrityl tetr 25g
SDP
Supplier:  Medchemexpress LLC 25GHYW127346
Encompass_Preferred
Pentaerythrityl tetrastearate is an ester compound commonly used as a lubricant emollient or thickener in various cosmetic and personal care products It is also used as a plasticizer or stabilizer in the production of plastics and resins Pentaerythrityl tetrastearate has unique chemical properties that make it an important ingredient in a variety of industrial processes including the manufacture of adhesives coatings and sealants
Catalog No. 50-002-05940