Learn More
Medchemexpress LLC Pentaethylene glycol | 4792-15-8 | 238.28 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0073411G
Encompass_Preferred
Pentaethylene glycol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. These PROTACs contain two different ligands connected by a linker, one designed for an E3 ubiquitin ligase and the other for the target protein. This mechanism allows PROTACs to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-60292