Learn More
Medchemexpress LLC Pentaethylene glycol | 4792-15-8 | 238.28 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0073415G
Encompass_Preferred
Pentaethylene glycol is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are molecules designed to degrade target proteins by recruiting E3 ubiquitin ligases. It appears as a colorless to light yellow liquid with a purity of 99.58%.
  • Used as a PROTAC linker
  • Enables synthesis of PROTACs
  • Facilitates targeted protein degradation
Catalog No. 50-004-60240