Learn More
Strem, An Ascensus Company CAS# 1765-40-8. 5g. Pentafluorobenzylbromide, 98+%. MFCD00000299
SDP
Supplier:  Strem, An Ascensus Company 0955505G
Encompass
CAS# 1765-40-8. 5g. Pentafluorobenzylbromide, 98+%. MFCD00000299. Molecular Weight: 260.99. Molecular Formula: C6F5CH2Br. Color/form: colorless liq. Strem# 09-5550. http://www.strem.com/catalog/v/09-5550/
Catalog No. 50-901-09370