Learn More
Strem, An Ascensus Company CAS# 35344-11-7. 1g. Pentamethylcyclopentadienyliron dicarbonyl dimer, min. 98%. MFCD00058862
SDP
Supplier:  Strem, An Ascensus Company 2635251G
Encompass
CAS# 35344-11-7. 1g. Pentamethylcyclopentadienyliron dicarbonyl dimer, min. 98%. MFCD00058862. Molecular Weight: 494.20. Molecular Formula: [(CH3)5C5Fe(CO)2]2. Color/form: red to purple xtl. Strem# 26-3525. http://www.strem.com/catalog/v/26-3525/
Catalog No. 50-901-11127