Learn More
Medchemexpress LLC Pentamethylcyclopentadienyl iridium(III) chloride dimer | 12354-84-6 | MFCD00075435 | 98.0% | 796.70 | C20H30Cl4Ir2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW01884550MG
Encompass_Preferred
[Ir(Cp-)Cl2]2 is an organometallic iridium(III) chloride dimer used as a precursor to catalysts for asymmetric transfer hydrogenation of ketones and in the synthesis of amines. It is supplied as an orange to reddish-brown solid for research use and requires light protection and inert storage.
  • Used as a precursor for asymmetric transfer hydrogenation catalysts.
  • Applicable in greener amine synthesis reactions.
  • Orange to reddish-brown solid, suitable for small-scale research.
  • Store at 4°C, protect from light, and keep under nitrogen for stability.
  • For research use only; not intended for human or patient use.
Catalog No. 50-003-00834