Learn More
Medchemexpress LLC Pentamidine | 100-33-4 | 93.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0537100MG
Encompass_Preferred
Pentamidine is an antimicrobial agent that interferes with DNA biosynthetics. It inhibits the parasite Leishmania infantum with an IC50 of 2.5 μM. It is also a potent and selective inhibitor of protein tyrosine phosphatases (PTPases) and phosphatase of regenerating liver (PRL). Pentamidine shows potential for the treatment of Gambian trypanosomiasis, antimony-resistant leishmaniasis, and Pneumocystis carinii pneumonia, and possesses antitumor and antibacterial activities.
  • Antimicrobial agent
  • Inhibits Leishmania infantum
  • Potent inhibitor of protein tyrosine phosphatases (PTPases) and phosphatase of regenerating liver (PRL)
  • Potential for treating Gambian trypanosomiasis, antimony-resistant leishmaniasis, and Pneumocystis carinii pneumonia
  • Exhibits antitumor and antibacterial activities
Catalog No. 50-004-08881