Learn More
Medchemexpress LLC Pentamidine | 100-33-4 | 97.2% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0537
Encompass_Preferred
Pentamidine is an antimicrobial agent that interferes with DNA biosynthesis. It is also a potent and selective inhibitor of protein tyrosine phosphatases (PTPases) and phosphatase of regenerating liver (PRL).
  • Antimicrobial agent
  • Interferes with DNA biosynthesis
  • Inhibits *Leishmania infantum*
  • Potent and selective inhibitor of protein tyrosine phosphatases (PTPases)
  • Potent and selective inhibitor of phosphatase of regenerating liver (PRL)
  • Potential for treating Gambian trypanosomiasis
  • Potential for treating antimony-resistant leishmaniasis
  • Potential for treating *Pneumocystis carinii* pneumonia
  • Exhibits antitumor activities
  • Exhibits antibacterial activities
Catalog No. 50-004-50704