Learn More
Medchemexpress LLC Pentamidine | 100-33-4 | 97.2% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB053750MG
Encompass_Preferred
Pentamidine is an antimicrobial agent that interferes with DNA biosynthesis. It inhibits the parasite Leishmania infantum and is a potent, selective inhibitor of protein tyrosine phosphatases (PTPases) and phosphatase of regenerating liver (PRL). It shows potential for treating Gambian trypanosomiasis, antimony-resistant leishmaniasis, and Pneumocystis carinii pneumonia, and also exhibits antitumor and antibacterial activities.
  • Interferes with DNA biosynthesis
  • Inhibits Leishmania infantum
  • Potent and selective inhibitor of PTPases and PRL
  • Potential for treating various parasitic infections
  • Exhibits antitumor and antibacterial activities
Catalog No. 50-004-05572