Learn More
TARGETMOL CHEMICALS INC Pentamidine 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T23133200MG
Encompass_Preferred
Also available in 5 mg, 500 mg and bulk. Please contact Fisher for quotes. Pentamidine is an antifungal and antiprotozoal agent, interacting directly with the pathogen genome by binding to AT-rich regions of duplex DNA and the minor groove of DNA, thereby interfering with DNA replication. Purity 98.13%
Catalog No. 50-287-7695