Learn More
Medchemexpress LLC Pentamidine isethionate | 140-64-7 | 99.95% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0537B100MG
Encompass_Preferred
Pentamidine isethionate (MP-601205 isethionate) is an antimicrobial agent that interferes with DNA biosynthetics. It inhibits the parasite *Leishmania infantum* with an IC50 of 2.5 μM. This compound is also a potent and selective inhibitor of protein tyrosine phosphatases (PTPases) and phosphatase of regenerating liver (PRL).
  • Antimicrobial agent
  • Interferes with DNA biosynthetics
  • Inhibits parasite *Leishmania infantum*
  • Potent and selective protein tyrosine phosphatases (PTPases) and phosphatase of regenerating liver (PRL) inhibitor
  • Potential applications in the treatment of Gambian trypanosomiasis, antimony-resistant leishmaniasis, and *Pneumocystis carinii* pneumonia
  • Demonstrates antitumor and antibacterial activities.
Catalog No. 50-004-08846