Learn More
Cayman Chemical PentaNacetylchItopentaose 5mg
SDP
Supplier:  Cayman Chemical 161015 MG
Encompass_Preferred
A pentameric chito-oligosaccharide involved in root nodulation; produced by NodC, a chito-oligosaccharide synthase, and serves as a substrate for NodL, an O-acetyltransferase; binds plant root lectins, an interaction that may be important in the promotion of cell division in the root cortex
Catalog No. 50-296-8841