Learn More
Medchemexpress LLC Pentanehydrazide (valeric acid hydrazide) | 38291-82-6 | ≥95.0% | 116.16 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01289010G
Encompass_Preferred
Pentanehydrazide (Valeric acid hydrazide) is a biochemical reagent that can function as an effective alkylating agent in the copper-catalyzed oxidative cyclization of N-arylacrylamides and participate in the tandem acylation reaction.
  • Biochemical reagent.
  • Acts as an effective alkylating agent in copper-catalyzed oxidative cyclization of N-arylacrylamides.
  • Participates in tandem acylation reactions.
Catalog No. 50-004-54894