Learn More
Medchemexpress LLC Pentanoic acid, 4-methyl-2-oxo-, sodium salt (1:1) | 4502-00-5 | MFCD00064193 | ≥98.0% | 152.12 | C6H9NaO3 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01738710G
Encompass_Preferred
Sodium α-ketoisocaproate is the sodium salt of α-ketoisocaproic acid (α-KIC), a metabolic intermediate of leucine used in biochemical and metabolic research. Supplied as a high-purity solid, it is commonly used to study insulin secretion, amino acid metabolism, and cellular energy pathways. The material is water-soluble and should be stored sealed and protected from moisture.
Catalog No. 50-002-99778