Learn More
Medchemexpress LLC Penthiopyrad | 183675-82-3 | 99.9% | 359.41 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1752010MM/1ML
Encompass_Preferred
Penthiopyrad (MTF-753) is a chiral carboxamide antifungal agent with a broad spectrum of fungicidal activity. It can be used for controlling foliar and soil-borne plant diseases on a broad range of agricultural crops and turfgrass. It is for research use only and not sold to patients.
  • Chiral carboxamide antifungal agent
  • Broad spectrum of fungicidal activity
  • Used for controlling foliar and soil-borne plant diseases
  • Applicable on a broad range of agricultural crops and turfgrass
  • Targets fungal pathogens
  • Acts via anti-infection pathway
Catalog No. 50-003-86325