Learn More
Medchemexpress LLC Penthiopyrad | 183675-82-3 | 99.9% | 359.41 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17520
Encompass_Preferred
Penthiopyrad is a chiral carboxamide antifungal agent with a broad spectrum of fungicidal activity. It can be used for controlling foliar and soil-borne plants diseases on a broad range of agricultural crops and turfgrass.
  • Chiral carboxamide antifungal agent.
  • Broad spectrum fungicidal activity.
  • Controls foliar plant diseases.
  • Controls soil-borne plant diseases.
  • Applicable on a broad range of agricultural crops.
  • Applicable on turfgrass.
Catalog No. 50-003-84289