Learn More
Medchemexpress LLC Pentosan Polysulfate (Sodium) | 140207-93-8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYA0203A5MG
Encompass_Preferred
Pentosan Polysulfate Sodium is an orally bioavailable, semi-synthetic medication with anti-inflammatory, pro-chondrogenic, and potent anti-HIV properties. It is used for the treatment of interstitial cystitis and is suitable for research applications.
  • Inhibits HIV-1 activity and antigen expression
  • Suppresses NF-κB and decreases proinflammatory actions of TNFα
  • Reduces MCP-1 production stimulated by high glucose and advanced glycation end products (AGEs)
  • Decreases interstitial inflammation and glomerulosclerosis in nephrectomized rats
  • Preserves renal function and reduces albuminuria
  • Mitigates severity of renal lesions, including tubulointerstitial inflammation
  • Reduces upregulation of TNFα and proinflammatory genes in aging diabetic kidneys
Catalog No. 50-004-32703