Learn More
TARGETMOL CHEMICALS INC Pentostatin 5mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T40065MG
Encompass_Preferred
Pentostatin is an extremely effective and irreversible inhibitor of adenosine deaminase (Ki: 2.5 pM).
Catalog No. NC1678286