Learn More
Cayman Chemical PentoxIfylIn 50g
SDP
Supplier:  Cayman Chemical 1872050 G
Encompass_Preferred
A methylxanthine derivative; has anti-inflammatory activity, inhibiting LPS-induced TNF-α production in isolated peripheral blood mononuclear cells (IC50 = 85 µM) and suppressing LPS-induced leukopenia in mice
Catalog No. 50-296-5058