Learn More
Medchemexpress LLC Pentoxyverine | 77-23-6 | 98.9% | 333.47 | C20H31NO3 | 5 MG

Supplier: Medchemexpress LLC HY1340045MG
Pentoxyverine is an antitussive and spasmolytic research chemical reported to act as a sigma-1 receptor agonist and muscarinic antagonist. It is supplied as a solid for biochemical and pharmacological studies and is used in receptor-binding and mechanism-of-action investigations.
- High purity suitable for analytical and pharmacological studies.
- Supplied as a solid in small milligram quantities.
- Useful for receptor binding and antitussive mechanism research.
- Molecular formula C20H31NO3 and molecular weight 333.47.
- CAS number 77-23-6 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.