Learn More
Cayman Chemical PentratIntrIfluoroacetate 10mg
SDP
Supplier:  Cayman Chemical 3687810 MG
Encompass_Preferred
A cell-penetrating peptide; has been used in the surface functionalization of liposomes for drug delivery across the BBB
Catalog No. 50-291-4900