Learn More
Cayman Chemical PeonIdInchlorIde 10mg
SDP
Supplier:  Cayman Chemical 1975410 MG
Encompass_Preferred
An O-methylated anthocyanidin that exhibits chemopreventive as well as anti-inflammatory activities on cancer cells in vitro, blocking COX-2 expression and transformation in JB6 P+ mouse epidermal cells
Catalog No. 50-298-2623