Learn More
Medchemexpress LLC Pepinh-MYD | 1421052-89-2 | 96.87% | 3388.03 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP107191MG
Encompass_Preferred
Pepinh-MYD is a MyD88 inhibitor that includes a domain sequence from MyD88 TIR and a protein transduction sequence, which allows it to penetrate the cell membrane. It disrupts MyD88-mediated TLR signaling pathways, thereby inhibiting associated immune responses. This compound has potential for studying the role of MyD88 in viral infections.
  • Penetrates cell membrane
  • Inhibits MyD88-mediated TLR signaling pathways
  • Potential for studying viral infections
Catalog No. 50-004-25798