Learn More
JPT PEPTIDE TECHNOLOGIES INC PEPMIX AAV9
Supplier:  JPT PEPTIDE TECHNOLOGIES INC PMAAV9VP1
Encompass
PepMix™ AAV9 (Capsid Protein VP1) Product Code: PM-AAV9-VP1 Protein ID: Q6JC40 Mix of 182 peptides
Catalog No. NC3911387