Learn More
Electron Microscopy Sciences Pepsin Reagent Ready To Use 15 ML
SDP
Supplier:  Electron Microscopy Sciences 6414205
Encompass_Preferred
Pepsin is used for proteolytic digestion of formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies. In IHC most commonly used fixative like formalin mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC. Pepsin digestion of FFPE tissue section improves accessibility of antibodies to tissue antigens.
Store in temperature of 2-8 degree C. DO NOT FREEZE.
Intended Use: Use as antigen retriever for some antibodies in IHC. Please refer to primary antibody protocol for IHC.
Catalog No. 50-192-6523