Learn More
TARGETMOL CHEMICALS INC PEPSTATIN 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T369550MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pepstatin (Pepsin Inhibitor S 735A) is a specific aspartic proteases inhibitor produced by actinomycetes and inhibits the aspartic proteases cathepsin D pepsin and renin. purity: 99%
Catalog No. 50-258-0157