Learn More
Gold Biotechnology Inc Pepstatin A 100 mg
SDP
Supplier:  Gold Biotechnology Inc P020100
Encompass_Preferred
Pepstatin A is a competitive, reversible aspartic protease inhibitor and inhibits proteases such as pepsin, chymosin, renin, HIV proteases, and cathepsins D and E. Peptstatin A is highly selective and is often used in a mixture with other enzyme inhibitors. In vitro studies have shown pepstatin A is the only aspartic acid inhibitor that exhibits inhibitory effects on HIV proteases.
Catalog No. NC2943376