Learn More
Selleck Chemical LLC Pepstatin A
SDP
Supplier:  Selleck Chemical LLC S7381200MG
Encompass_Preferred
Pepstatin A (Pepstatin) is a potent aspartic protease inhibitor and also inhibits HIV replication Pepstatin A is also an inhibitor of cathepsins D and cathepsins E Pepstatin A inhibits autophagy by suppressing lysosomal proteases
Catalog No. 50-000-04434