Learn More
Abcam Pepstatin A, aspartic protease inhibitor, 100MG
Supplier:  Abcam AB141416100MG
Encompass_Preferred
MW 685.9 Da, Purity >90%. Pepstatin A, aspartic protease inhibitor. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6636