Learn More
Carbosynth LLC. Pepstatin A (Synthetic), 100mg, MW: 685.91, Molecular formula: C34H63N5O9, CAS# 26305-03-3
SDP
Supplier:  Carbosynth LLC. IPA3998PI100MG
Encompass_Preferred
Synonyms: Iva-Val-Val-Sta-Ala-Sta-OH, Isovaleryl-L-Valyl-L-Valyl[(3S,4S)-4-Amino-3-Hydroxy-6-Methylheptanoyl]-L-Alanyl[(3S,4S)-4-Amino-3-Hydroxy-6-Methylheptanoic Acid], Application: Synthetic Product Inhibitor for Pepsin, Cathepsin D/E and Renin, Storage Temperature: -20°C
Catalog No. 50-168-5887